Learn More
ACROBiosystems Biotinylated Human CD27 Ligand / CD70 Protein, His, Avitag™
Biotinylated Protein
Marke:  ACROBiosystems CDL-H82Q9-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16407399
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Cluster of Differentiation 70 (CD70) is also known as CD27 ligand (CD27L / CD27LG), TNFSF7, TNFSF7G, is a type II transmembrane glycoprotein belonging to the TNF superfamily (TNFSF) and is a surface antigen found on activated T-and B-lymphocytes and mature dendritic cells. Binding of CD70 to its receptor CD27 induces in priming, effector functions, differentiation and memory formation of T-cells, and thus is invloved in the biological processes including T-cell activation, the proliferation of costimulated T-cells, as well as the generation of cytolytic T-cells. CD70 on T cells provides costimulatory signals that are required for T cell proliferation, clonal expansion and the promotion of effector T cell formation. CD70 on mouse B cell has been shown to inhibit terminal differentiation of activated B cells into plasma cells and enhances commitment to memory B cell responses. CD70 induces proliferation and IFNγ production, on NK cells.
This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.7 kDa. The protein migrates as 27-37 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001243.1
25 μg
Gln 39 - Pro 193
CD70,CD27LG,TNFSF7,TNFSF7G,CD27L
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
20.7 kDa
Biotinylated Human CD27 Ligand, His,Avitag (CDL-H82Q9) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Pro 193 (Accession # P32970-1).
-20°C
CD27 Ligand
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins