Learn More
ACROBiosystems CD47: SIRP alpha, Biotinylated, Inhibitor Screening ELISA Assay Pair
This pair is useful for screening for inhibitors of human CD47 binding to human SIRP alpha
Marke:  ACROBiosystems EP-102-96tests
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16407499
  • 605.28 € / 96 Tests
Voraussichtliches Versanddatum: 30-08-2021
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Immune checkpoint pathway is a focal point of today’s cancer research. CD47 is one of the best characterized checkpoint proteins. CD47 binding to SIRP alpha aids in tumor evasion of the immune system. Therefore, the pharmaceutical inhibition of CD47 has been considered a promising strategy by many oncologists.
Spezifikation
Spezifikation
Inhibitory Kit
-20°C, aviod light
RT
This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new CD47 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human CD47 and in
96 Tests