Learn More
ACROBiosystems Human ErbB3 / Her3 Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems ER3-H5257-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ErbB3,also known as Her3 (human epidermal growth factor receptor 3), is a member of the epidermal growth factor receptor (EGFR) family of receptor tyrosine kinases. This membrane-bound glycoprotein has a neuregulin binding domain but has not an active kinase domain. It therefore can bind the ligand but cannot mediate the intracellular signal transduction through protein phosphorylation. However, it does form heterodimers with ErbB2 or other EGFR members responsible for tyrosine phosphorylation to give a receptor complex and initiate the related pathway, which lead to cell proliferation or differentiation. Overexpression of this protein has been reported in numerous cancers, including prostate, bladder, and breast tumors. This protein has different isoforms derived from alternative splicing variants, and among which, the secreted isoform lacking the intermembrane region modulates the activity of membrane-bound form.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 95.5 kDa. The protein migrates as 115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001973.2 | |
100 μg | |
Ser 20 - Thr 643 | |
ERBB3,HER3,LCCS2,MDA-BF-1,MGC88033,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
95.5 kDa | |
Human ErbB3, Mouse IgG2a Fc Tag (ER3-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1). | |
-20°C | |
ErbB3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |