Learn More
ACROBiosystems Human Integrin alpha 4 beta 1 (ITGA4&ITGB1) Heterodimer Protein, His Tag&Tag Free
Marke:  ACROBiosystems IT1-H52W1-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16407789
  • 3838.91 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Integrins are transmembrane proteins that mediate interactions between adhesion molecules on adjacent cells and/or the extracellular matrix (ECM). Integrins have diverse roles in several biological processes including cell migration during development and wound healing, cell differentiation, and apoptosis. Their activities can also regulate the metastatic and invasive potential of tumor cells. Integrin alpha 4 beta 1(Alpha-4/beta-1) is receptors for fibronectin. Integrin alpha-4/beta-1 is a receptor for VCAM1 and recognizes the sequence Q-I-D-S in VCAM1.
Human ITGA4&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA4 and ITGB1, has a calculated MW of 111.4 kDa (ITGA4) and 83.7 kDa (ITGB1). Subunit ITGA4 is fused with an acidic tail at the C-terminus and followed by a polyhistid
Spezifikation
Spezifikation
NP 000876.3 (ITGA4) and NP_002202.2 (ITGB1)
1 mg
Tyr 34 - Thr 977 (ITGA4) and Gln 21 - Asp 728 (ITGB1)
Integrin alpha 4 beta 1,ITGA4andITGB1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
111.4 kDa (ITGA4) and 83.7 kDa (ITGB1)
Human ITGA4&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W1) is expressed from human 293 cells (HEK293). It contains AA Tyr 34 - Thr 977 (ITGA4) and Gln 21 - Asp 728 (ITGB1) (Accession # P13612-1 (ITGA4) and P05556-1 (ITGB1)).
-20°C
Integrin alpha 4 beta 1
Human
50 mM Tris, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens