missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus Siglec-9 Protein, His Tag

Marke:  ACROBiosystems SI9-C52H9-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16408069

  • 353.08 € / 100 Mikrogramm

Voraussichtliches Versanddatum: 19-10-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

siglec-9 (HGMW-approved symbol SIGLEC9) a member of the sialic acid-binding Ig-like lectin (Siglec) family, which belongs to the immunoglobulin superfamily (IgSF). SIGLEC9 shows a high degree of homology to many members of the siglec family, including siglec-7 (80%), siglec-8 (72%), siglec-5 (65%), and CD33 (64%). This high degree of homology is also conserved in the extracellular Ig-like domains. They are characterized by an N-terminal Ig-like V-type domain which mediates sialic acid binding, followed by varying numbers of Ig-like C2-type domains。Siglec-9 with a hydrophobic signal peptide, an N-terminal Ig-likeV-type domain, two Ig-like C2-type domains, a transmembrane region and a cytoplasmic tail.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.0 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

XP_005590140.1
HEK293
Cynomolgus
38.0 kDa
100 µg
Gln 20 - Gly 350
Siglec-9,SIGLEC9,CDw329,CD329
Unconjugated
25 mM MES, 150 mM NaCl, pH5.5
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Cynomolgus Siglec-9, His Tag (SI9-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 350 (Accession # XP_005590140.1).
-20°C
Siglec-9
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate