Learn More
ACROBiosystems Canine TIGIT Protein, His Tag
Marke: ACROBiosystems TIT-C52H7-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TIGIT (also called T cell immunoreceptor with Ig and ITIM domains) is one newly discovered immune receptor on some percentage of T cells and Natural Killer Cells(NK). It is also identified as WUCAM and Vstm3. TIGIT could bind to CD155(PVR) on dendritic cells(DCs), macrophages, etc. with high affinity, and also to CD112(PVRL2) with lower affinity. TIGIT expressing CD8+ T cells has been shown to be expanded and associated with clinical markers of HIV disease progression in a diverse group of HIV infected individuals. TIGIT and PD-1 has been shown to be over expressed on tumor antigen-sepcific (TA-specific) CD8+ T cells and CD8+ tumor infiltrating lymphocytes (TILs) from individuals with melanoma. Blockade of TIGIT and PD-1 led to increased cell proliferation, cytokine production, and degranulation of TA-specific CD8+ T cells and TIL CD8+ T cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
23.1 kDa | |
Canine TIGIT, His Tag (TIT-C52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 208 (Accession # F1PKJ0-1). | |
-20°C | |
TIGIT | |
Canine | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
1 mg | |
Phe 17 - Pro 208 | |
TIGIT,VSIG9,VSTM3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |