Learn More
ACROBiosystems Biotinylated Human BCMA / TNFRSF17 Protein, His, Avitag™ DMF Filed
Biotinylated Protein
Marke: ACROBiosystems BCA-H82E4-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 12 kDa and 15-17 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001183.2 | |
200 μg | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
9.6 kDa | |
Biotinylated Human BCMA, His,Avitag (BCA-H82E4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). | |
-20°C | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |