Learn More
ACROBiosystems Biotinylated Human BTN3A2 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems BT2-H82E7-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Butyrophilin subfamily 3 member A2(BTN3A2/BT3.2) mRNA expression by tumoral cells was previously identified as a prognostic factor in a small cohort of high grade serous epithelial ovarian cancer (HG-EOC). We find that the three BTN3A isoforms: BTN3A1, BTN3A2, and BTN3A3, have high structural homology to the B7 superfamily of proteins and exist as V-shaped homodimers in solution, associating through the membrane proximal C-type Ig domain.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.3 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001184175.1 | |
25 μg | |
Gln 30 - Trp 248 | |
BT3.2,BT3.3,BTF3,BTF4 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
27.3 kDa | |
Biotinylated Human BTN3A2, His,Avitag (BT2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # P78410-1). | |
-20°C | |
BTN3A2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |