Learn More
ACROBiosystems Human CD94 Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems CD4-H5253-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD94(Natural killer cells antigen CD94) is also known as KLRD1, KP43, NK cell receptor, Killer cell lectin-like receptor subfamily D member 1. CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
44.0 kDa | |
Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1). | |
-20°C | |
CD94 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Lys 32 - Ile 179 | |
CD94,KLRD1,KP43,NK cell receptor | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |