Learn More
ACROBiosystems Mouse CD24 Protein, His Tag
Marke: ACROBiosystems CD4-M52H7-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 4.6 kDa. The protein migrates as 20-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_033976.1 | |
100 μg | |
Asn 27 - Gly 53 | |
CD24,CD24A | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
4.6 kDa | |
Mouse CD24, His Tag (CD4-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Gly 53 (Accession # P24807-1). | |
-20°C | |
CD24 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |