missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human Fc gamma RIIB / CD32b Protein, Avitag™,His Tag (SPR & BLI verified)

Biotinylated Protein

Marke:  ACROBiosystems CDB-H82E0-200ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16417359

  • 1392.13 € / 200 Mikrogramm

Auf Lager, versandfertig: 1

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. Low affinity immunoglobulin gamma Fc region receptor II-b (FCGR2B) is also known as CD32b, FCG2, IGFR2. CD32B is expressed on B cells and myeloid dendritic cells. Ligation of CD32B on B cells downregulates antibody production and may, in some circumstances, promote apoptosis. Co-ligation of CD32B on dendritic cells inhibits maturation and blocks cell activation. CD32B may also be a target for monoclonal antibody therapy for malignancies.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 23.0 kDa. The protein migrates as 25 kDa and 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_003992.3
HEK293
Human
23.0 kDa
200 µg
Ala 46 - Pro 217
FCGR2B,CD32b,CD32,FCG2,IGFR2,CDw32
Precision Avi
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human CD32b, Avitag,His Tag (SPR and BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994-1).
-20°C
Fc gamma RIIB / CD32b
His Tag and Avi Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate