Learn More
ACROBiosystems Biotinylated Human CEACAM-8 / CD66b Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems CE8-H82E9-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Description
Carcinoembryonic antigen-related cell adhesion molecule 8 (CEACAM8) is also known as CD66b (Cluster of Differentiation 66b), CD66b, CD67, CGM6, NCA-95, and is one of seven human CEACAM family members within the immunoglobulin superfamily. CEACAM family members are a set of widely expressed proteins involved in several biological functions, including cell adhesion, migration, signal transduction, and the regulation of gene expression. Abnormal overexpression and downregulation of some CEACAMs have been described in tumor cells. In humans, CEACAMs include type I transmembrane proteins (CEACAM1, CEACAM3, and CEACAM4) and GPI-linked molecules (CEACAM5 through CEACAM8). There is no human CEACAM2. CEACAM8 is a single chain, two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is an activation marker for human granulocytes.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 35.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001807.2 | |
200 μg | |
Gln 35 - Ser 319 | |
CEACAM8,CD66b,CD67,CGM6,NCA-95 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
35.1 kDa | |
Biotinylated Human CEACAM-8, His,Avitag (CE8-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession # P31997-1). | |
-20°C | |
CEACAM-8 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |