Learn More
ACROBiosystems Biotinylated Bovine FcRn / FCGRT&B2M Heterodimer Protein, His, Avitag™
Biotinylated SPR verified Protein
Marke:  ACROBiosystems FCN-B82W3-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16417529
  • 1847.47 € / 200 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
FCGRT & B2M heterodimer protein (FcRn complex) consist of two subunits: p51 (equivalent to FCGRT), and p14 (equivalent to beta-2-microglobulin), and forms an MHC class I-like heterodimer. Fc fragment of IgG, receptor, transporter, alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Biotinylated Bovine FcRn Heterodimer Protein, His,Avitag (SPR verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.5 kDa (FCGRT) and 11.6 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus, followed by
Spezifikation
Spezifikation
NP_788830.1 (FCGRT) and NP_776318.1 (B2M)
200 μg
Ala 24 - Pro 298 (FCGRT) and Ile 21 - Leu 118 (B2M)
FcRn,FCGRT and B2M
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
34.5 kDa (FCGRT) and 11.6 kDa (B2M)
MABSol™ Biotinylated Bovine FcRn Heterodimer Protein, His,Avitag (SPR verified) (FCN-B82W3) is expressed from human HEK293 cells. It contains AA Ala 24 - Pro 298 (FCGRT) and Ile 21 - Leu 118 (B2M) (Accession # Q3T119-1 (FCGRT) and P01888-1 (B2M)).
-20°C
FcRn (FCGRT and B2M)
Bovine
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins