Learn More
ACROBiosystems Human Integrin alpha 11 beta 1 (ITGA11&ITGB1) Heterodimer Protein, His Tag&Tag Free
Marke: ACROBiosystems IT1-H52W3-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Integrin alpha 11 beta 1 is a receptor for collagen. The alpha11 cDNA encodes a mature protein with a large 1120-residue extracellular domain that contains an I-domain of 207 residues and is linked by a transmembrane domain to a short cytoplasmic domain of 24 amino acids. The deduced alpha11 protein shows the typical structural features of integrin alpha-subunits and is similar to a distinct group of alpha-subunits from collagen-binding integrins.
Human ITGA11&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA11 and ITGB1, has a calculated MW of 131.6 kDa (ITGA11) and 82.7 kDa (ITGB1). Subunit ITGA11 is fused with an acidic tail at the C-terminus and followed by a polyhiSpezifikation
NP_001004439.1 (L524R) (ITGA11) and NP_002202.2 (ITGB1) | |
100 μg | |
Phe 23 - Pro 1141 (ITGA11) and Gln 21-Asp 728 (ITGB1) | |
Integrin alpha 11 beta 1,ITGA11andITGB1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
131.6 kDa (ITGA11) and 82.7 kDa (ITGB1) | |
Human ITGA11&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Pro 1141 (ITGA11) and Gln 21-Asp 728 (ITGB1) (Accession # Q9UK x 5-1 (L524R) (ITGA11) and P05556-1 (ITGB1)). | |
-20°C | |
Integrin alpha 11 beta 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |