Learn More
ACROBiosystems Biotinylated Human Prolactin / PRL Protein, Fc, Avitag™
Biotinylated Protein
Marke:  ACROBiosystems PRN-H82F7-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16417979
  • 1847.47 € / 200 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Prolactin (gene name PRL) is a secreted neuroendocrine pituitary hormone that acts primarily on the mammary gland to promote lactation, but has pleiotropic effects in both males and females. Prolactin is synthesized as a prohormone. Following cleavage of the signal peptide, the length of the mature hormone is between 194 and 199 amino acids, depending on species. Hormone structure is stabilized by three intramolecular disulfide bonds. Excessive secretion of prolactin - hyperprolactinemia - is a relative common disorder in humans. This condition has numerous causes, including prolactin-secreting tumors and therapy with certain drugs. The prolactin receptor (gene name PRLR) is a transmembrane type I glycoprotein that belongs to the cytokine hematopoietic receptor family. Expression of the prolactin receptor is widespread. Each prolactin molecule is thought to bind two receptor molecules
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.2 kDa. The protein migrates as 57-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_000939.1
200 μg
Leu 29 - Cys 227
PRL,Prolactin
Precision Avi
Fc Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
51.2 kDa
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Accession # Q5THQ0-1).
-20°C
Prolactin
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins