Learn More
ACROBiosystems Cynomolgus Siglec-15 / CD33L3 Protein, Fc Tag
Marke: ACROBiosystems SG5-C5253-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Siglec-15 is a DAP12-associated immunoreceptor, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Siglecs are cell surface proteins that bind sialic acid. They are found primarily on the surface of immune cells and are a subset of the I-type lectins. Siglec-15 consisting of immunoglobulin (Ig)-like domains, transmembrane domain and a short cytoplasmic tail. Siglec-15 is that recognizes sialylated glycans and regulates osteoclast differentiation. Siglec-15 is a potential therapeutic target for osteoporosis and plays a conserved regulatory role in the immune system of vertebrates.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
52.7 kDa | |
Cynomolgus Siglec-15, Fc Tag (SG5-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # A0A2K5UY47-1). | |
-20°C | |
Siglec-15 | |
Cynomolgus Monkey | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
100 μg | |
Phe 20 - Thr 263 | |
CD33 antigen-like 3,SIGLEC-15,CD33L3,sialic acid-binding Ig-like lectin 15,Siglec15,Siglec-15 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |