Learn More
ACROBiosystems Biotinylated Human Siglec-15 / CD33L3 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems SG5-H82F5-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Siglec-15 is a DAP12-associated immunoreceptor, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Siglecs are cell surface proteins that bind sialic acid. They are found primarily on the surface of immune cells and are a subset of the I-type lectins. Siglec-15 consisting of immunoglobulin (Ig)-like domains, transmembrane domain and a short cytoplasmic tail. Siglec-15 is that recognizes sialylated glycans and regulates osteoclast differentiation. Siglec-15 is a potential therapeutic target for osteoporosis and plays a conserved regulatory role in the immune system of vertebrates.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.8 kDa. The protein migrates as 58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_998767.1 | |
200 μg | |
Phe 20 - Thr 263 | |
CD33 antigen-like 3,SIGLEC-15,CD33L3,sialic acid-binding Ig-like lectin 15,Siglec15,Siglec-15 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
54.8 kDa | |
Biotinylated Human Siglec-15, Fc,Avitag (SG5-H82F5) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # Q6ZMC9-1). | |
-20°C | |
Siglec-15 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |