Learn More
ACROBiosystems Rhesus macaque SLAMF7 / CRACC / CD319 Protein, His Tag
Marke: ACROBiosystems SL7-R52H7-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
SLAM family member 7 (SLAMF7) is also known as CD2-like receptor-activating cytotoxic cells (CRACC), Membrane protein FOAP-12, CD antigen CD319, Novel Ly9, Protein 19A, which is a single-pass type I membrane protein and a member of the CD2 family of cell surface receptors. SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Isoform 1 of SLAMF7 mediates NK cell activation through a SH2D1A-independent extracellular signal-regulated ERK-mediated pathway. May play a role in lymphocyte adhesion. Isoform 3 of SLAMF7 does not mediate any NK cell activation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 35-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_001117618.1 | |
100 μg | |
Ser 23 - Met 226 | |
SLAMF7,CD319,CS1,CRACC,19 A,FOAP-12 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
24.4 kDa | |
Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # F7HQ72-1). | |
-20°C | |
SLAMF7 | |
Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |