Learn More
ACROBiosystems Biotinylated Human BTLA (31-150) Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems BTA-H82E6-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 28-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAP44003.1 | |
200 μg | |
Lys 31 - Ser 150 | |
BTLA,CD272 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
17.4 kDa | |
Biotinylated Human BTLA (31-150), His,Avitag (BTA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1). | |
-20°C | |
BTLA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |