Learn More
ACROBiosystems Biotinylated Human CD52 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems CD2-H82E8-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CAMPATH-1 antigen, also known as cluster of differentiation 52 (CD52), is a glycoprotein that in humans is encoded by the CD52 gene.It is widely expressed on the cell surface of immune cells, such as mature lymphocytes, natural killer cells (NK), eosinophils, neutrophils, monocytes/macrophages, and dendritic cells (DCs).ligation of cell surface CD52 molecules may offer costimulatory signals for T-cell activation and proliferation.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 13.5 kDa. The protein migrates as 16 kDa and 17-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001794.2 | |
200 μg | |
Gly 25 - Ser 36 | |
CDW52,HE5,CD52,CAMPATH-1 antigen | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
13.5 kDa | |
Biotinylated Human CD52, His,Avitag (CD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1). | |
-20°C | |
CD52 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |