missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus IL-3 R alpha / CD123 Protein, His Tag

SPR verified

Marke:  ACROBiosystems ILA-C52H6-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16427719

  • 423.38 € / 100 Mikrogramm
Verfügbar ab: 24-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

34.7 kDa
Cynomolgus IL-3 R alpha, His Tag (SPR verified) (ILA-C52H6) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession # G8F3K3-1).
-20°C
IL-3 R alpha
Cynomolgus Monkey
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
100 μg
Arg 18 - Arg 302
IL3R,IL3RA,IL-3 Ra,IL-3 R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3 Ra,MGC34174,IL-3 R alpha
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote