Learn More
ACROBiosystems Human IL-12 R beta 1 / CD212 Protein, His Tag
Marke: ACROBiosystems ILB-H52H9-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The human IL-12 R subunit is a member of the cytokine receptor superfamily and the functional high-affinity IL-12R is composed of at least two beta-type cytokine receptor subunits, each independently exhibiting a low affinity for IL-12. IL-12 R beta 1 (Interleukin-12 receptor subunit beta-1) is also known as IL-12RB1, CD212. Functions as an interleukin receptor which binds interleukin-12 with low affinity and is involved in IL12 transduction. Associated with IL12RB2 it forms a functional, high affinity receptor for IL12. Associates also with IL23R to form the interleukin-23 receptor which functions in IL23 signal transduction probably through activation of the Jak-Stat signaling cascade.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.0 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_005526.1 | |
100 μg | |
Cys 24 - Glu 540 | |
IL-12 R beta 1,IL12RB1,CD212,IL12R,IL12RB,IL-12 RB1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
59.0 kDa | |
Human IL-12 R beta 1, His Tag (ILB-H52H9) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession # P42701-1). | |
-20°C | |
IL-12 R beta 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |