Learn More
ACROBiosystems FITC-Labeled Human Mesothelin / MSLN (296-580) Protein, Fc Tag
Marke: ACROBiosystems MSN-HF25x-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 59.1 kDa. The protein migrates as 60-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH09272.1 | |
25 μg | |
Glu 296 - Gly 580 | |
MSLN,Mesothelin,MPF | |
FITC | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
59.1 kDa | |
FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF25x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H526x). | |
-20°C, avoid light | |
Mesothelin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |