Learn More
ACROBiosystems Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag™&Tag Free
MALS verified
Marke: ACROBiosystems CDD-C82W6-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 17.8 kDa (CD3E) and 13.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag and followed by an Avi tagSpezifikation
NP_001270544.1 (CD3E) and NP_001274617.1 (CD3D) | |
25 μg | |
Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
17.8 kDa (CD3E) and 13.5 kDa (CD3D) | |
Biotinylated Cynomolgus CD3E&CD3D; Heterodimer Protein, His,Avitag™&Tag; Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) and Q95LI8-1 (CD3D)). | |
-20°C | |
CD3E and CD3D | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |