Learn More
ACROBiosystems Human GITR Ligand / TNFSF18 Protein, His Tag
Marke: ACROBiosystems GIL-H5249-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor ligand superfamily member 18 (TNFSF18) is also known as Glucocorticoid - induced TNF-related ligand (GITRL or GITR Ligand), Activation - inducible TNF - related ligand (AITRL), which belongs to the tumor necrosis factor family. TNFSF18 is expressed at high levels in the small intestine, ovary, testis, kidney and endothelial cells. TNFSF18 / GITRL is up-regulated after stimulation by bacterial lipopolysaccharides (LPS). TNFSF18 Can function as costimulator and lower the threshold for T-cell activation and T-cell proliferation. TNFSF18 / GITR Ligand is important for interactions between activated T-lymphocytes and endothelial cells. TNFSF18 also mediates activation of NF-kappa-B.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.5 kDa. The protein migrates as 23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
20.5 kDa | |
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession # AAH69319.1). | |
-20°C | |
GITR Ligand | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
100 μg | |
Gln 50 - Ser 177 | |
TNFSF18,AITRL,TL6,hGITRL,GITR Ligand | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |