Learn More
ACROBiosystems Mouse Serum Albumin Protein, His Tag
Marke: ACROBiosystems MSA-M52H8-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
serum albumin (SA) is also known as ALB, which is the main protein of plasma and has a good binding capacity for water,Ca2+,Na+,K+,fatty acids,hormones, bilirubin and drugs.The main function of SA is the regulation of the colloidal osmotic pressure of blood. As Major zinc transporter in plasma, SA typically binds about 80% of all plasma zinc. A variant structure of albumin could lead to increased binding of zinc resulting in an asymptomatic augmentation of zinc concentration in the blood. Defects in serum albumin can cause familial dysalbuminemic hyperthyroxinemia which is a form of euthyroid hyperthyroxinemia that is due to increased affinity of serum albumin for T4. It is the most common cause of inherited euthyroid hyperthyroxinemia in Caucasian population.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.8 kDa. The protein migrates as 65-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_033784.2 | |
1 mg | |
Glu 25 - Ala 608 | |
Serum albumin,ALB,Alb | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
67.8 kDa | |
Mouse Serum Albumin, His Tag (MSA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1). | |
-20°C | |
Serum Albumin | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |