Learn More
ACROBiosystems Human TrkA / NTRK1 (192-402) Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems TRA-H5253-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Receptor tyrosine kinase involved in the development and the maturation of the central and peripheral nervous systems through regulation of proliferation, differentiation and survival of sympathetic and nervous neurons. High affinity receptor for NGF which is its primary ligand. Can also bind and be activated by NTF3/neurotrophin-3. However, NTF3 only supports axonal extension through NTRK1 but has no effect on neuron survival. Upon dimeric NGF ligand-binding, undergoes homodimerization, autophosphorylation and activation. Recruits, phosphorylates and/or activates several downstream effectors including SHC1, FRS2, SH2B1, SH2B2 and PLCG1 that regulate distinct overlapping signaling cascades driving cell survival and differentiation. In absence of ligand and activation, may promote cell death, making the survival of neurons dependent on trophic factors.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001012331.1 | |
100 μg | |
Gly 192 - Val 402 | |
TrkA,NTRK1,MTC,TRK,TRKA,Trk-A | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
49.1 kDa | |
Human TrkA (192-402), Mouse IgG2a Fc Tag (TRA-H5253) is expressed from human 293 cells (HEK293). It contains AA Gly 192 - Val 402 (Accession # P04629-2). | |
-20°C | |
TrkA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |