Learn More
ACROBiosystems Human IL-3 R alpha / CD123 Protein, His Tag (MALS & SPR verified)
Marke: ACROBiosystems ILA-H52H6-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002174.1 | |
100 μg | |
Thr 19 - Arg 305 | |
IL3R,IL3RA,IL-3 Ra,IL-3 R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3 Ra,MGC34174,IL-3 R alpha | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
35.0 kDa | |
Human IL-3 R alpha, His Tag (ILA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1). | |
-20°C | |
IL-3 R alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |