Learn More
ACROBiosystems Rhesus macaque / Cynomolgus Complement Factor D / CFD Protein, His Tag (active enzyme)
Marke: ACROBiosystems CFD-R52H3-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Complement factor D (CFD) is also known as Adipsin, C3 convertase activator, Properdin factor D (PFD), which contains one peptidase S1 domain and belongs to the peptidase S1 family. CFD / Adipsin cleaves factor B when the latter is complexed with factor C3b, activating the C3bbb complex, which then becomes the C3 convertase of the alternate pathway. CFD / Adipsin is a serine protease that stimulates glucose transport for triglyceride accumulation in fats cells and inhibits lipolysis. Defects in CFD / Adipsin are the cause of complement factor D deficiency which predisposes to invasive meningococcal disease.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 25-27 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_005587397.1 | |
100 μg | |
Ile 26 - Ala 253 | |
CFD,Adipsin,PFD,DF,Complement factor D | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
26.5 kDa | |
Rhesus macaque / Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Ala 253 (Accession # H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Complement Factor D are homologus. | |
-20°C | |
Complement Factor D | |
Rhesus Macaque, Cynomolgus Monkey | |
50 mM Tris, 150 mM NaCl, pH8.0 | |
HEK293 | |
Enzymes |