missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus / Rhesus macaque FGL1 Protein, Fc Tag

Marke:  ACROBiosystems FG1-C5269-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16457529

  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Fibrinogen-like protein 1(FGL1) is also known as HP-041, Hepassocin, HFREP-1, LFIRE-1. The protective effect of fibrinogen-like protein 1 (FGL1) in liver injury has previously been reported. However, studies have shown that FGL1 may be a predictor of GC patients and a target for GC therapy. Immunocytochemical studies revealed that fgl1 selectively binds to defective spermatozoa in the cauda epididymidis. Northern blot analysis and in situ hybridization demonstrated the high expression of fgl1 in the principal cells of the proximal cauda epididymidis. Immunofluorescence analysis using mouse fibrotic lung tissues suggested that fibrotic regions showed increased expressions of Gtse1 and Fgl1, Gtse1 and Fgl1 are suggested to be novel targets for radiation-induced lung fibrosis.

This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

60.4 kDa
Cynomolgus / Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # G7N0K6-1).
-20°C
FGL1
Cynomolgus Monkey, Rhesus Macaque
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens
100 μg
Leu 23 - Ile 312
FGL1,Hepassocin,HP-041,HFREP-1,LFIRE-1,HFREP1
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote