Learn More
ACROBiosystems Biotinylated Human ICOS / CD278 (C136S, C137S) Protein, His,Avitag™
Recommended for biopanning
Marke: ACROBiosystems ICS-H82E5-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Inducible T-cell costimulator (ICOS) is also known as Activation-inducible lymphocyte immunomediatory molecule (AILIM), CD278, which belongs to the CD28 family of immune costimulatory receptors consisting of CD28, CTLA-4 and PD-1. ICOS enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells. CD278 / ICOS prevents the apoptosis of pre-activated T-cells and also plays a critical role in CD40-mediated class switching of immunoglobin isotypes.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_036224.1 (C136S, C137S) | |
200 μg | |
Glu 21 - Phe 141 | |
ICOS,CD278,AILIM,Inducible T-cell costimulator | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
17.4 kDa | |
Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Phe 141 (Accession # Q9Y6W8-1 (C136S, C137S)). | |
-20°C | |
ICOS | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |