Learn More
ACROBiosystems Biotinylated Human Mucin-1 (890-1158) Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems MU1-H82E5-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Mucin-1 (MUC1) is also known as Tumor-associated epithelial membrane antigen (EMA), Polymorphic epithelial mucin (PEM), Peanut-reactive urinary mucin (PUM), PEMT, Krebs von den Lungen-6 (KL-6), CD antigen CD227, Episialin, H23AG. MUC1 is a glycoprotein with extensive O-linked glycosylation of its extracellular domain. Mucins line the apical surface of epithelial cells in the lungs, stomach, intestines, eyes and several other organs. Mucins protect the body from infection by pathogen binding to oligosaccharides in the extracellular domain, preventing the pathogen from reaching the cell surface. Except protective function by binding to pathogens, MUC1 also functions in a cell signaling capacity. Overexpression of MUC1 is often associated with colon, breast, ovarian, lung and pancreatic cancers.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag.The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has aSpezifikation
21.3 kDa (partial α chain) and 10.2 kDa (partial β chain) | |
Biotinylated Human Mucin-1 (890-1158), His,Avitag (MU1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ser 890 - Gly 1158 (Accession # P15941-1). | |
-20°C | |
Mucin-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
200 μg | |
Ser 890 - Gly 1158 | |
Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1/SEC,MUC-1/X,MUC1/ZD,PEM,PEMT,PUM,CA 15-3,Episialin | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |