Learn More
ACROBiosystems Human NKG2A / CD159a Protein, Fc Tag
Brand: ACROBiosystems NKA-H526c-100ug
Additional Details : Netto-Gewicht : 0.00010kg
Description
NKG2A/CD159a is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors that inhibits innate immune system activation, also known as KLRC1, CD159a, NK cell receptor A and NKG2-A/NKG2-B type II integral membrane protein. NKG2A marks a unique immune effector subset preferentially co-expressing the tissue-resident CD103 molecule, but not immune checkpoint inhibitors. NKG2A blockade therapy operated through CD8 T cells, but not NK cells. The increase in NKG2A expression might be induced by IL-10, which was present at a high level in the plasma of HCC patients. Blocking IL-10 could specifically inhibit NKG2A expression in NK cells. These findings indicate that NKG2A expression is influenced by factors from cancer nests and contributes to NK cell exhaustion, suggesting that NKG2A blockade has the potential to restore immunity against liver tumors by reversing NK cell exhaustion.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_002250.1 | |
100 μg | |
Pro 94 - Leu 233 | |
NKG2A,CD159a,KLRC1,NK cell receptor A | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.5 kDa | |
Human NKG2A, Fc Tag (NKA-H526c) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # P26715-1). | |
-20°C | |
NKG2A | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |