Learn More
ACROBiosystems Human CD48 / SLAMF2 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems BC1-H5258-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD48 antigen is also known as Signaling lymphocytic activation molecule 2, SLAM family member 2 (SLAMF2), Leukocyte antigen MEM-102, BCM1 surface antigen, B-lymphocyte activation marker BLAST-1. CD48 is a 65 kDa GPI-linked protein which belongs to the CD2 subfamily of immunoglobulin superfamily molecules. CD48 antigen contains two Ig-like C2-type (immunoglobulin-like) domains and two Ig-like V-type (immunoglobulin-like) domain. CD48 interacts with CD244 in an heterophilic manner. CD48 is ligand for CD2. CD48 might facilitate interaction between activated lymphocytes. CD48 is probably involved in regulating T-cell activation.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 64-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001769.2 | |
100 μg | |
Gln 27 - Ser 220 | |
CD48,BCM1,SLAMF2,BLAST,BLAST1,MEM-102,TCT0.1,BCM-1,SLAMF-2,BLAST-1 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
49.2 kDa | |
Human CD48, Mouse IgG2a Fc Tag, low endotoxin (BC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # P09326-1). | |
-20°C | |
CD48 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |