Learn More
ACROBiosystems FITC-Labeled Human CD30 / TNFRSF8 Protein, His Tag
Marke:  ACROBiosystems CD0-HF2H3-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16467259
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Human CD30 is also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. TNFRSF-8 is expressed by activated, but not by resting, T and B cells. Also, CD30 is expressed on classical Hodgkin Lymphoma cells together with CD15. CD30 is the receptor for TNFSF8/CD30L. CD30 can interact with TRAF2 and TRAF5, and mediate the signal transduction that leads to the activation of NF-kappa-B. TNFRSF8 may play a role in the regulation of cellular growth and transformation of activated lymphoblasts. TNFRSF8 is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.3 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001234.2
25 µg
Phe 19 - Lys 379
TNFRSF8,CD30,D1S166E,Ki-1
FITC
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>90% as determined by SDS-PAGE
39.3 kDa
FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF2H3) is expressed from human HEK293 cells. It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). It is the FITC labeled form of Human CD30, His Tag (Cat. No. CD0-H5229).
-20°C, avoid light
CD30
Human
PBS, pH7.4
HEK293
Fluorescent labeled protein