Learn More
ACROBiosystems Biotinylated Cynomolgus Serum Albumin Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems HSA-C82E5-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
serum albumin (SA) is also known as ALB, which is the main protein of plasma and has a good binding capacity for water,Ca2+,Na+,K+,fatty acids,hormones, bilirubin and drugs.The main function of SA is the regulation of the colloidal osmotic pressure of blood. As Major zinc transporter in plasma, SA typically binds about 80% of all plasma zinc. A variant structure of albumin could lead to increased binding of zinc resulting in an asymptomatic augmentation of zinc concentration in the blood. Defects in serum albumin can cause familial dysalbuminemic hyperthyroxinemia which is a form of euthyroid hyperthyroxinemia that is due to increased affinity of serum albumin for T4. It is the most common cause of inherited euthyroid hyperthyroxinemia in Caucasian population.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 69.7 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001306284.1 | |
25 µg | |
Asp 25 - Ala 608 | |
Serum albumin,ALB,Alb | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
69.7 kDa | |
Biotinylated Cynomolgus Serum Albumin, His,Avitag (HSA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ala 608 (Accession # A2V9Z4-1). | |
-20°C | |
Serum Albumin | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |