Learn More
ACROBiosystems Biotinylated Human IL-2 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems IL2-H82F3-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interleukin-2 (IL-2) is an interleukin, a type of cytokine immune system signaling molecule, which is a leukocytotrophic hormone that is instrumental in the body's natural response to microbial infection and in discriminating between foreign (non-self) and self. IL-2 mediates its effects by binding to IL-2 receptors, which are expressed by lymphocytes, the cells that are responsible for immunity. Mature human IL-2 shares 56% and 66% aa sequence identity with mouse and rat IL-2, respectively. Human and mouse IL-2 exhibit crossspecies activity. The receptor for IL-2 consists of three subunits that are present on the cell surface in varying preformed complexes. IL-2 is also necessary during T cell development in the thymus for the maturation of a unique subset of T cells that are termed regulatory T cells (T-regs). After exiting from the thymus, T-Regs function to prevent other T cells from recognizing and reacting against “self antigens”, which could result in “autoimmunity”. T-Regs do so by preventing the responding cells from producing IL-2. Thus, IL-2 is required to discriminate between self and non-self, another one of the unique characteristics of the immune system.
This protein carries a human IgG1 Fc tag at the N-terminus and an Avi tag at the C-terminus. The protein has a calculated MW of 43.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000577.2 | |
25 μg | |
Ala 21 - Thr 153 | |
IL2,TCGF,lymphokine,Interleukin 2 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
43.7 kDa | |
Biotinylated Human IL-2, Fc,Avitag (IL2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P60568-1). | |
-20°C | |
IL-2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |