missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human PD-1 / PDCD1 Protein, Mouse IgG2a Fc Tag

HPLC-verified

Marke:  ACROBiosystems PD1-H5255-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16467949

  • 302.64 € / 100 Mikrogramm

Voraussichtliches Versanddatum: 19-10-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.

This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_005009.2
HEK293
Human
42.5 kDa
100 µg
Leu 25 - Gln 167
PDCD1,PD1,CD279,SLEB2
Unconjugated
PBS, pH7.4
Less than 0.1 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (PD1-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).
-20°C
PD-1
Mouse IgG Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate