Learn More
ACROBiosystems Unconjugated Human TPBG / 5T4 Protein, His, Avitag™
Unconjugated Protein
Marke: ACROBiosystems TPG-H52E5-50ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Trophoblast glycoprotein (TPBG), also known as 5T4, is the therapeutic target of several anticancer agents currently in clinical development, largely due to its high expression in tumors and low expression in normal adult tissues.This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. TPBG is expressed by all types of trophoblasts as early as 9 weeks of development.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.0 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001159864.1 | |
50 μg | |
Ser 32 - Ser 355 | |
TPBG,5T4,M6P1,5T4AG,WAIF1,5T4 oncofetal antigen,Trophoblast glycoprotein,5T4 oncofetal trophoblast glycoprotein | |
Unconjugated | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
39.0 kDa | |
Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession # Q13641-1). | |
-20°C | |
TPBG | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |