Learn More
ACROBiosystems Human CD7 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems CD7-H5258-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16477319
  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
T-cell antigen CD7 (CD7) is also known as GP40, LEU-9, TP41 and Tp40. CD7 is a protein that in humans is encoded by the CD7 gene, this gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily. CD7 has been shown to interact with PIK3R1. This protein is found on thymocytes and mature T cells. It plays an essential role in T-cell interactions and also in T-cell/B-cell interaction during early lymphoid development.
This protein carries a llama IgG2b Fc tag at the N-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_006128.1
100 μg
Ala 26 - Pro 180
CD7,GP40,TP41,LEU-9,Tp40
Unconjugated
Llama IgG Fc Tag
Less than 0.01 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
44.3 kDa
Human CD7, Llama IgG2b Fc Tag, low endotoxin (CD7-H5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).
-20°C
CD7
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens