Learn More
ACROBiosystems Human Serum Albumin Protein, His Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems HSA-H522a-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Artikelnummer. 16477639
  • 539.84 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
serum albumin (SA) is also known as ALB, which is the main protein of plasma and has a good binding capacity for water,Ca2+,Na+,K+,fatty acids,hormones, bilirubin and drugs.The main function of SA is the regulation of the colloidal osmotic pressure of blood. As Major zinc transporter in plasma, SA typically binds about 80% of all plasma zinc. A variant structure of albumin could lead to increased binding of zinc resulting in an asymptomatic augmentation of zinc concentration in the blood. Defects in serum albumin can cause familial dysalbuminemic hyperthyroxinemia which is a form of euthyroid hyperthyroxinemia that is due to increased affinity of serum albumin for T4. It is the most common cause of inherited euthyroid hyperthyroxinemia in Caucasian population.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.4 kDa. The protein migrates as 67-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_000468.1
1 mg
Asp 25 - Leu 609
Serum albumin,ALB,Alb
Unconjugated
His Tag
Less than 0.01 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
67.4 kDa
Human Serum Albumin, His Tag, low endotoxin (HSA-H522a) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # P02768-1).
-20°C
Serum Albumin
Human
PBS, pH7.4
HEK293
Other Recombinant Proteins