Learn More
ACROBiosystems Human CLEC12A / MICL / CLL-1 Protein, His Tag
Marke: ACROBiosystems CLA-H5245-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CLEC12A (C-type lectin domain family 12 member A) is also known as CLL1, DCAL2, MICL. Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death. Cell surface receptor that modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases. Evidence of distinct disease propagating stem cells in myelodysplastic syndrome (MDS) has emerged in recent years. The role of CLEC12A in MDS, however, remains to be elucidated. Furthermore, CLEC12A has been proposed as a promising marker of leukaemic stem cells in AML.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_612210.4 | |
1 mg | |
His 65 - Ala 265 | |
CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
25.6 kDa | |
Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2). | |
-20°C | |
CLEC12A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |