Learn More
ACROBiosystems Human GFR alpha-like Protein, His Tag
Marke: ACROBiosystems GFA-H52H3-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GFR alpha-like is also known as GDNF family receptor alpha-like, GFRAL, C6orf144, UNQ9356, PRO34128. Growth differentiation factor-15 (GDF15) is a circulating protein that has been implicated in multiple biological processes, including energy homeostasis, body weight regulation, and cachexia driven by cancer and chronic disease. GDNF family receptor α-like (GFRAL) was recently identified as the neuronal brainstem receptor responsible for mediating the anorectic actions of GDF15. Brainstem-restricted receptor for GDF15 which regulates food intake, energy expenditure and body weight in response to metabolic and toxin-induced stresses . Upon interaction with its ligand, GDF15, interacts with RET and induces cellular signaling through activation of MAPK- and AKT- signaling pathways.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_997293.2 | |
100 μg | |
Ser 19 - Glu 351 | |
GFR alpha like,GFR alpha-like,GFRAL,GRAL | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.7 kDa | |
Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession # Q6UXV0-1). | |
-20°C | |
GFR alpha-like | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |