Learn More
ACROBiosystems Rat IGF-I R / CD221 Protein, His Tag
Marke:  ACROBiosystems IGR-R5224-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16487669
  • 3083.12 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The Insulin-like Growth Factor 1 Receptor (IGF1) is also known as CD221, JTK13. and is a transmembrane receptor that is activated by IGF-1 and by the related growth factor IGF-2. It belongs to the large class of tyrosine kinase receptors. This receptor mediates the effects of IGF-1, which is a polypeptide protein hormone similar in molecular structure to insulin. IGF1R is make up of two alpha subunits and two beta subunits ,the Both the α and β subunits are synthesized from a single mRNA precursor. The precursor is then glycosylated, proteolytically cleaved, and crosslinked by cysteine bonds to form a functional transmembrane αβ chain.The α chains are located extracellularly while the β subunit spans the membrane and are responsible for intracellular signal transduction upon ligand stimulation. IGF1R have a binding site for ATP, which is used to provide the phosphates for autophosphorylation. There is a 60% homology between IGF1R and the insulin receptor. In response to ligand binding, the α chains induce the tyrosine autophosphorylation of the β chains. This event triggers a cascade of intracellular signaling that, while somewhat cell type specific, often promotes cell survival and cell proliferation.
This protein carries a polyhistidine tag at the C-terminus.This protein contains a furin convertase cleavage site, 738-RRRR-741, and will be partially processed into N (α chain) and C-terminal fragment (partial β chain) with calculated MW of 81.3 kDa and
Spezifikation
Spezifikation
NP_434694.1
1 mg
Glu 31 - His 936
IGF1R,IGFR,JTK13,CD221,MGC142170,MGC142172,MGC18216
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
81.3 kDa and 24.2 kDa
Rat IGF-I R, His Tag (IGR-R5224) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # P24062-1).
-20°C
IGF-I R
Rat
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens