Learn More
ACROBiosystems Human IL-17 RE (155-454) Protein, Fc Tag
Marque:  ACROBiosystems ILE-H5256-100ug
Informations supplémentaires : Netto-Gewicht : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16487759
  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description
Description
Interleukin-17 receptor E (IL-17 RE) is an orphan receptor of the IL-17 receptor family, also known as UNQ3056, PRO9877, IL-17 Receptor E. IL-17RE is known to be expressed in the pancreas, brain, and prostate. IL-17RE is a receptor specific to IL-17C and has an essential role in host mucosal defense against infection. IL-17C functioned in a unique autocrine manner, binding to a receptor complex consisting of the receptors IL-17RA and IL-17RE, which was preferentially expressed on tissue epithelial cells. Some data identify IL-17RE as a receptor of IL-17C that regulates early innate immunity to intestinal pathogens.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
NP_705613.1
100 μg
Thr 155 - His 454
IL-17 RE,IL17RE,IL-17 RE,IL-17 receptor E
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
60.5 kDa
Human IL-17 RE (155-454), Fc Tag (ILE-H5256) is expressed from human 293 cells (HEK293). It contains AA Thr 155 - His 454 (Accession # Q8NFR9-1).
-20°C
IL-17 RE
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications