Learn More
ACROBiosystems Human LIGHT / TNFSF14 Protein, His Tag
Marke: ACROBiosystems LIT-H5247-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor ligand superfamily member 14(LIGHT) is a tumor necrosis factor (TNF) superfamily ligand that regulates T cell immune responses by signaling through the herpes virus entry mediator (HVEM) and the lymphotoxin beta receptor (LTbetaR). LIGHT has emerged as a potent initiator of T cell co-stimulation signals effecting CTL-mediated tumor rejection, allograft rejection and graft versus host disease. Constitutive expression of LIGHT leads to tissue destruction and autoimmune-like disease syndromes.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 24-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP 003798.2 | |
100 µg | |
Asp 74 - Val 240 | |
CD258 antigen,CD258,HVEM-L,LIGHT,LTg,TNFSF14,TR2,HVEML | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
20.9 kDa | |
Human LIGHT, His Tag (LIT-H5247) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1). | |
-20°C | |
LIGHT | |
Human | |
PBS with Arginine, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |