Learn More
ACROBiosystems Biotinylated Human TPBG / 5T4 Protein, His, Avitag™
Biotinylated Protein
Marke:  ACROBiosystems TPG-H82Eb-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16488109
  • 443.87 € / 25 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Trophoblast glycoprotein (TPBG), also known as 5T4, is the therapeutic target of several anticancer agents currently in clinical development, largely due to its high expression in tumors and low expression in normal adult tissues.This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. TPBG is expressed by all types of trophoblasts as early as 9 weeks of development.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.0 kDa. The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_006661.1
25 μg
Ser 32 - Ser 355
TPBG,5T4,M6P1,5T4AG,WAIF1,5T4 oncofetal antigen,Trophoblast glycoprotein,5T4 oncofetal trophoblast glycoprotein
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
39.0 kDa
Biotinylated Human TPBG, His,Avitag (TPG-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession # Q13641-1).
-20°C
TPBG
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins