Learn More
ACROBiosystems Human CD206 / MMR Protein, His Tag
Marke: ACROBiosystems CD6-H52H9-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Mrc1 is a conserved checkpoint mediator protein that transduces the replication stress signal to the downstream effector kinase. Mrc1 and its vertebrate homologue Claspin serve as a mediator for replication stress checkpoint signaling, receiving the signal from Mec1/Rad3/ATR sensor kinase and transmitting it to the effector Rad53/Cds1/Chk1 kinase. The loss of mrc1 checkpoint activity results in the aberrant activation of late/dormant origins in the presence of hydroxyurea. Tumor-associated macrophages (TAMs) expressing the multi-ligand endocytic receptor mannose receptor (CD206/MRC1) contribute to tumor immunosuppression, angiogenesis, metastasis, and relapse.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 158.6 kDa. The protein migrates as 150-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002429.1 | |
100 μg | |
Leu 19 - Ala 1389 | |
MMR,CD206,hMR,MRC1,CLEC13D,CLEC13DL,MRC1L1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
158.6 kDa | |
Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession # P22897-1). | |
-20°C | |
CD206 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |