Learn More
ACROBiosystems Human DLL3 Protein, Fc Tag
Marke: ACROBiosystems DL3-H5255-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.0 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_058637.1 | |
100 µg | |
Ala 27 - Leu 492 | |
Delta3,delta-like 3 (Drosophila),delta-like protein 3,DLL3,Pudgy,SCDO1,SCDO1delta3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
75.0 kDa | |
Human DLL3, Fc Tag (DL3-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1). | |
-20°C | |
DLL3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |