Learn More
ACROBiosystems Mouse Flt-3 / Flk-2 Protein, His Tag
Marke: ACROBiosystems FL3-M52H6-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Flt-3(Receptor-type tyrosine-protein kinase FLT3) is also known as FLK-2(Fetal liver kinase-2), STK-1(Stem cell tyrosine kinase 1), CD135. FLT3 is a cytokine receptor which belongs to the receptor tyrosine kinase class III. Tyrosine-protein kinase that acts as cell-surface receptor for the cytokine FLT3LG and regulates differentiation, proliferation and survival of hematopoietic progenitor cells and of dendritic cells. Promotes phosphorylation of SHC1 and AKT1, and activation of the downstream effector MTOR. Promotes activation of RAS signaling and phosphorylation of downstream kinases, including MAPK1/ERK2 and/or MAPK3/ERK1. Mutations that cause constitutive kinase activity promote cell proliferation and resistance to apoptosis via the activation of multiple signaling pathways.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.9 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_034359.2 | |
100 μg | |
Asn 28 - Ser 544 | |
Flt-3,Flk-2,STK-1,CD135,FLK2,FLT-3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
59.9 kDa | |
Mouse Flt-3, His Tag (FL3-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 28 - Ser 544 (Accession # NP_034359.2). | |
-20°C | |
Flt-3 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |